Learn More
Thermo Scientific™ Forma™ HEPA2 VOC Filter
Catalog No. 11688022 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
HEPA2 VOC Filter
Catalog No. Type
11-688-022 HEPA2 VOC Filter
1 options
Catalog No. 11-688-022 Supplier Thermo Scientific™ Supplier No. 760200
Add to Cart
Add to Cart
For Use With: Forma Steri-cycle and Forma series II Water jacketed CO2 Incubator
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
  • Precise control
  • Choice of TC or IR sensors
  • Superior parameter recovery characteristics
  • Continuous contamination control technology
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Forma™ Steri-Cycle™, Direct Heat and Water Jacket
Type HEPA2 VOC Filter