Description
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
- Precise control
- Choice of TC or IR sensors
- Superior parameter recovery characteristics
- Continuous contamination control technology
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| CO2 Sensor Technology | TC Sensor or IR Sensor |
| For Use With (Equipment) | Forma Series 310 Direct heat CO2 incubator |
| Lighting | LED |
| Type | HEPA Filter |