Learn More
Thermo Scientific™ Wash Buffer for GeneJET Gel Extraction Kit
Description
Thermo Scientific™ GeneJET Wash Buffer is a component of the GeneJET Gel Extraction Kit (Cat No. K0691, K0692) and may be purchased separately. Provided in 45 mL aliquots.
Specifications
Specifications
| Chemical Name or Material | Wash Buffer |
| For Use With (Equipment) | GeneJET Gel Extraction Kit |
| Product Line | GeneJET |
| Quantity | 45 mL |
| Solution Type | Buffer |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.