Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Cylindrical Longpath Cell Holder
Catalog No. 14385450 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cylindrical Cell Holder
Catalog No. Type
14-385-450 Cylindrical Cell Holder
1 options
Catalog No. 14-385-450 Supplier Thermo Scientific™ Supplier No. 4011000
Add to Cart
Add to Cart
Cylindrical Longpath Cell Holder. Snap-in” holder accepts one cylindrical longpath cell up to 50mm long and 22mm in diameter.
TRUSTED_SUSTAINABILITY
Specifications
Type Cylindrical Cell Holder
For Use With (Equipment) Genesys™ 20 spectrophotometer
Pathlength Up to 50 mm