Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Rectangular Longpath Cell Holder
Catalog No. 14385451 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cell Holder
Catalog No. Type
14-385-451 Cell Holder
1 options
Catalog No. 14-385-451 Supplier Thermo Scientific™ Supplier No. 4012000
Add to Cart
Add to Cart
Rectangular Longpath Cell Holder. Easy-to-install “snap-in” cell holder accommodates one rectangular longpath cell up to 50mm long and 12mm wide.
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Holder
For Use With (Equipment) Genesys™ 20 spectrophotometer
Holds Cell up to 50m L and 12mm W
Pathlength 50 mm
Shape Rectangular