Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Single-position Cell Holder
Shop All Thermo Scientific ProductsDescription
Single-position Cell Holder. As supplied with GENESYS 20 Spectrophotometer. Holds one 10mm square cuvette, 1/2 in. or 10mm diameter test tube.
Specifications
Specifications
| Type | Cell Holder |
| For Use With (Equipment) | Genesys™ 20 spectrophotometer |
| Holds | One 10 mm square cuvette, 0.5 in. or 10 mm dia. test tube |
| Pathlength | 10 mm |