Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation and Temperature Map Documention
Shop All Thermo Scientific ProductsDescription
IQ/OQ/Temperature Mapping documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific ULT Freezers: Forma 88000-series, HeraFreeze HFUT, Thermo Scientific TSU, Revco UxF, Thermo Scientific TSX (pre 2018) |
| Description | Documentation Only for the following, Services Not included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. Temperature Mapping: 24 hours of multi-point mapping. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ/Temp Map Documentation |