Learn More
Thermo Scientific™ HAAKE™ Viscotester™ iQ Rheometer Application Packages
Catalog No. 2620020 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Liquids
Liquids, Pastes
Yield Point Determination
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
26-200-20 Yield Point Determination
26-200-05 Liquids
26-200-10 Liquids, Pastes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 26-200-20 Supplier Thermo Scientific™ Supplier No. 2620020
Add to Cart
Add to Cart
Request a Quote
Ready to use in seconds in any configuration you need
Acquire simple viscosity curves or perform complex rheological investigations covering measurements of low viscous liquids up to highly viscous pastes with the Thermo Scientific™ HAAKE™ Viscotester™ rheometer, which can be used as a standalone unit or optionally, be controlled by software. The size and footprint of the HAAKE™ Viscotester™ rheometer make it a portable unit suitable for mobile use or allow it to serve as an important tool in the quality control laboratory.
Application packages are designed to perform a diverse set of rheological tasks and meet a variety of application needs.
  • Advanced design with one-hand lift operation
  • Touchscreen control panel with multilingual, graphical user interface
  • Data quality assessment and QC quality criteria
  • Connect Assist quick coupling and automatic recognition of temperature modules and rotors
  • Liquid temperature module uses a circulator for coaxial cylinders and parallel plates
  • Self-sufficient Peltier temperature module for coaxial cylinders and parallel plates
  • Small dimensioned, coaxial cylinders use less sample and shorter temperature equilibration times to minimize costs per measurement
  • Universal container holder for relative measurements in the original containers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Rheometer Application Package
Includes HAAKE Viscotester iQ Rheometer, Universal container holder for original containers and Rotor FL22 4B/SS
For Use With (Application) Yield Point Determination