Learn More
Thermo Scientific™ HBB-6/HLR-6 Bucket Adapters
Catalog No. 11358 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
Triple, Quint or Quad Bags
Catalog No. Capacity
11-358 Triple, Quint or Quad Bags
1 options
Catalog No. 11-358 Supplier Thermo Scientific™ Supplier No. 11358
Add to Cart
Add to Cart
Enable blood bag centrifuging in the Sorvall RC3BP
For use in the HBB-6/HLR-6 rotors.
  • Adapters accept triple, quint or quad blood bags
  • Maximum capacity: 6 x 550mL
TRUSTED_SUSTAINABILITY
Specifications
Capacity Triple, Quint or Quad Bags
For Use With (Equipment) HLR-6 bucket for swing bucket rotor