Learn More
Epredia™ HiPur™ Methyl Alcohol
Catalog No. 22110887 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
5 gallon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-110-887 5 gallon
22-110-675 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-110-887 Supplier Epredia™ Supplier No. 96005
Add to Cart
Add to Cart
  • Anhydrous
  • Meets spectrophotometric grade specifications
  • Final filtration is 0.2μm
Specifications
Product Type Alcohol
Quantity 5 gallon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov