Learn More
Thermo Scientific™ HyperSep™ Retain PEP SPE Columns
Catalog No. 60107222 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
10 mg
Catalog No. Bed Weight
60-107-222 10 mg
1 options
Catalog No. 60-107-222 Supplier Thermo Scientific™ Supplier No. 60107222
Versatile polymeric material for retention of polar and non-polar analytes
  • Exceptional recoveries for polar and nonpolar analytes
  • High and consistent recoveries
  • High capacity material is a high purity, highly porous polystyrene DVB material modified with urea functional groups to give balanced retention of polar and non-polar analytes
  • Fast and easy sample preparation and method development
  • pH stable 0 to 14
Drugs and metabolites in biological fluids; Peptides in serum, plasma or biological fluids; Environmental samples
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 10 mg