Learn More
Unity Lab Services™ Thermo Scientific™ Laboratory Refrigerator Validation and Temperature Map Documention for Thermo Scientific™ TSX505
Catalog No. IOPQDE03813 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific TSX505 Laboratory Refrigerator
Catalog No. For Use With (Equipment)
IOPQDE03813 Thermo Scientific TSX505 Laboratory Refrigerator
1 options
Catalog No. IOPQDE03813 Supplier Unity Lab Services™ Supplier No. IOPQDOCE89003813
Add to Cart
Add to Cart
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Laboratory Refrigerator Validation and Temperature Map Documention for Thermo Scientific TSX505
IQ/OQ/Temperature Mapping documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific TSX505 Laboratory Refrigerator
Description Documentation Only for the following, Services Not included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. Temperature Mapping: 24 hours of multi-point mapping.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ/Temp Map Documentation