Learn More
Description
- Double injection molded for maximum sample delivery and best fit for a variety of pipetters
- Available in 200 and 1000μL including bulk and racked formats
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Volume (Metric) | 0.5 to 200 μL |
| Color | Yellow |
| Autoclavable | Autoclavable |
| Format | Bulk |
| For Use With (Equipment) | LabTip Pipette |
| Product Line | LabTip |
| Material | Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

