Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtips Pipette Tips
GREENER_CHOICE

Catalog No. p-3207576
Change view
Click to view available options
Volume (Metric):
0.5 to 200 μL
1000 μL
Color:
Blue
Yellow
Format:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Format
50101183 1000 μL Blue Rack
50101182 0.5 to 200 μL Yellow Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50101183 Supplier Thermo Scientific™ Supplier No. 94300230
Only null left
Add to Cart
Add to Cart

Produced from the highest quality virgin polypropylene

  • Double injection molded for maximum sample delivery and best fit for a variety of pipetters
  • Available in 200 and 1000μL including bulk and racked formats

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Format Rack
Tip Style Universal Fit
For Use With (Equipment) LabTip Pipette
Product Line LabTip
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.