Description
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators.
Specifications
Specifications
| Description | Reinforced Floor with Removable Ramp |
| For Use With (Equipment) | Model 3948 and 3962 Environmental Chamber/Incubator |
| Lighting | LED |
| Type | Reinforced Floor Option |
| Installation | Customer installed |