Learn More
Unity Lab Services™ Thermo Scientific Liquid Nitrogen Storage Equipment Validation and Temperature Map Documention for Thermo Scientific CryoPlus™ Storage Systems
Shop All Thermo Scientific ProductsDescription
IQ/OQ/Temperature Mapping documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoPlus™ Storage Systems |
| Description | Documentation Only for the following, Services Not included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. Temperature Mapping: 24 hours of multi-point mapping. Includes Freezing Cycle testing of liquid phase cooling , super cooling, phase change to solid. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ/Temp Map Documentation |