Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix Mobile ARIS Detection System

Catalog No. 18001139 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectors:
Matrix Mobile ARIS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectors
18-001-139 Matrix Mobile ARIS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Products on this page may include voltages not considered standard in North America. Please review carefully.
Catalog No. 18-001-139 Supplier Thermo Scientific™ Supplier No. SYSA001000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Products on this page may include voltages not considered standard in North America. Please review carefully.

Specifications

Detectors Matrix Mobile ARIS
For Use With (Application) For fast isotope identification, correlates GPS coordinates with survey data for comparison during patrols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.