Learn More
Thermo Scientific™ MIC 2000 Microtiter microplate
GSA_VAGREENER_CHOICE
Catalog No. 1424590 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
14-245-90 Sterile
1 options
Catalog No. 14-245-90 Supplier Thermo Scientific™ Supplier No. 120151
Add to Cart
Add to Cart
For support of microbiolgical applications
These products are suitable for applications including bacterial growth, bacterial identification, minimum inhibitory concentration (MIC), hybridization, hybridoma plating and cloning.
  • Microtiter microplate format
  • Polystyrene
  • U-bottom
Order Info
Bag of 20; Box of 100
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.28 mL
Well Shape Round Bottom (U-well)