Learn More
Thermo Scientific™ Microtiter™ Immulon™ 384 Square Well Microplates
Shop All Thermo Scientific ProductsDescription
Ideal for high-throughput screening and assay miniaturization.
- Standard microplate footprint
- Choice of universal and enhanced surfaces
- Well shape: Rounded square
- Well bottom: Flat-bottom well
- Well volume: 120 μL
Surface
- Immulon 1 B - Hydrophobic surface for preferential binding of biomolecules with hydrophobic domains
- Immulon 4 HBX - Hydrophilic surface designed for high binding of IgG as well as a very wide range of proteins and biomolecules that have hydrophilic/hydrophobic character
Specifications
Specifications
| Volume (Metric) | 120 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Format | Rack |
| For Use With (Equipment) | Thermo Scientific Matrix |
| Product Line | Matrix |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.