Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mobile Detection System

Catalog No. 19175632
Change view
Click to view available options
Energy Range:
36 keV to 1.3 MeV
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Energy Range
19-175-632 36 keV to 1.3 MeV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-175-632 Supplier Thermo Scientific™ Supplier No. 4255013

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mobile radiation reconnaissance system designed to operate in a vehicle for use on the ground or in airspace

System is used to discover/map the location of artificial radiation sources or survey large areas of contamination. Rugged, lightweight, modular radiation detection system that can easily be deployed in any type of moving vehicle to search for unusual or abnormal radioactivity. It can also be used to survey large contaminated areas, by air, to discover localized hot spots and map the event. The Mobile Detection System (MDS) is comprised of a large volume, highly sensitive organic plastic scintillator detector, coupled to a global positioning based movable mapping system, that automatically tracks both the position and corresponding radiation measurement of the local background as the vehicle performs its mission. The data are stored to a .dbf file which can be easily exported to other programs or viewed later from the exclusive MapTrack PC program.

  • Mobile gamma-/neutron-radiation reconnaissance system operating in vehicle or helicopter (aircraft)
  • Rugged, easily relocated system with user friendly features
  • The MDS-GN has data transmission capability for radio modem or GSM cellular devices
  • Allows for a wide range of users without compromising accuracy and results
  • Remote viewing and/or control of mission from command station via wireless transmission of data
  • Fully integrated ArcView GIS for simultaneous hazard modeling and verification with real time data
  • Lightweight, modular design allows for easy relocation between vehicles
  • No hard wiring into the vehicle is necessary
  • Real time data feed into any version
  • ESRI ArcView software is standard
  • battery provides 8 hours of continuous use when fully charged
  • Can also be deployed in helicopters, emergency vehicles
  • Utilizes Natural Background Rejection (NBR) technology for instant threat assessment of radiological incidents
  • Comes with Zargus military grade carrying case
MapTrack
  • MapTrack program is a global positioning based movable mapping system that automatically records radiation measurement data along the path of travel.
  • Utilizes a 6 channel Global Positioning System (GPS) to closely track the exact coordinates.
  • MAPTRACK software (included) provides a real-time data feed into any standard version of ESRIs ArcView/ ArcGIS software (ArcView 3.3, ArcGIS versions 8 and 9)
  • MAPTRACK and any version of ArcView can be used separately or simultaneously
Specifications
  • Detector Type: Gamma/neutron radiation
  • Voltage: 12 VDC
  • Detection Range: 1nSv/Hour to 20μSv/hr. Scintillation detector, 10nSv/hr. to 1Sv/hr. proportional detector
  • Weight: 15kg (33.1 lb.)
  • Humidity Range: 5 to 95%
  • Operating Temperature: 0 to 50°C
  • Test Sensitivity: 20000cps/μSv/hr. to 137 Cs Scintillation detector, 10cps/μS/hr. neutron detector
  • Energy Range: 36keV to 1.3MeV

TRUSTED_SUSTAINABILITY

Specifications

Detector Type Gamma /Neutron Radiation
Sensitivity 20000 cps/μSv/Hour to 137 Cs Scintillation detector, 10cps/μS/Hour neutron detector
Voltage 12 VDC
Weight (English) 33.1 lb.
Weight (Metric) 15 kg
Energy Range 36 keV to 1.3 MeV
Range 1 nSv/Hour to 20 μSv/Hour Scintillation detector, 10 nSv/Hour to 1 Sv/Hour proportional detector
Humidity Range 5% to 95%
Temperature (Metric) Operating 0°C to 50°C
Product Type Mobile Detection System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.