Learn More
Thermo Scientific™ Multi™ and Multi RF™ Centrifuges Eight-Place Fixed-Angle Rotor
Shop All Thermo Scientific ProductsDescription
Includes
Rotor and aerosol-tight lid
Ideal for removing insoluble material, protein and peptide recovery, cell lysates and oligos concentration.
- Capacity: 8 × 50mL
- Maximum speed/RCF: 15,100rpm/25,200xG
- Directly accepts up to 8 × 50mL tubes, conical-bottom BD Falcon™ or Corning™ tubes
- Angle: 25°; radius 9.9cm
Order Info
Adapters (05-375-20, available online) reduce capacity to accept up to 8 x 15mL Falcon or Corning conicals.
Specifications
Specifications
| Type | Rotor |
| Capacity | 8 x 50 mL |
| Max. RCF | 25,200 x g |
| Max. Speed | 15,100 rpm |
| Max. Tube Length | 120 mm |
| Max. Tube Diameter | 17 mm |
| For Use With (Equipment) | Thermo Scientific IEC Centrifuge |
| Angle | Fixed |
| Includes | Lids aerosol containment |
| No. of Places | 8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.