Learn More
Cole-Parmer Thermo Scientific Nalgene 2158-0021 Quick Filling/Venting Closures, PP, 83b 2-port 1/4"
Catalog No. NC1262202 Shop All Cole-Parmer Products
Encompass
Ideal for filling/dispensing into and out of carboys and bottles with an 83B neck finish. Quick disconnect tubing ports make installation a snap. Order replacement straight hose barb inserts and optional elbow hose barb inserts below. Note: Polyproplene is autoclavable.
Catalog No. NC1262202 Supplier Cole-Parmer Supplier No. 6250800
Ideal for filling/dispensing into and out of carboys and bottles with an 83B neck finish. Quick disconnect tubing ports make installation a snap. Order replacement straight hose barb inserts and optional elbow hose barb inserts below. Note: Polyproplene is autoclavable.