Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System

Catalog No. 1256518
Click to view available options
Wells:
1
2
4
8
16
Material:
Permanox Plastic
Soda Lime Glass
This item is not returnable. View return policy

Catalog No. 12-565-18

Supplier: Thermo Scientific™ 177402

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Grow anchorage dependent cells right on the microscope slide using the unique removable chamber of the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System.

Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. Slide surface is specially treated for adherent cell culture. Choose either specialty washed Soda Lime Glass slide or cell attachment optimized Permanox Plastic slide. Media chamber is easily removable without the need for a tool.

  • Broad range of formats and well number to meet application needs
  • Available with a Permanox solvent-resistant plastic slide for adherent cell culture
  • Writing surface provides easy identification of samples
  • Save time and reagents by growing cells directly on microscope slide
  • In 1, 2, 4, 8 and 16 well configurations; all measure 25 × 75mm
  • Medical-grade silicone sealing gasket can be removed for cover slipping
  • No cell transfer needed prior to visualization/staining
  • Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
  • Suitable for use with fluorescent labels
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Wells 8
Culture Area 0.8 cm2
Lid Material Polystyrene
Description 8-well Chamber Slide w/ removable wells
Surface Treatment Cell Culture
Material Soda Lime Glass
Volume (Metric) Working 0.2 to 0.4 mL
Includes Clear polystyrene lid with handle on the side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 75 x 25 mm
Sterility Sterile
Closure Color Clear
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.