Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ 3-Star Benchtop Application Kit: pH of Hair Care Products

Catalog No. 13643068 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Measures hair care product pH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
13-643-068 Measures hair care product pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-643-068 Supplier Thermo Scientific™ Supplier No. 1010160

Includes

Meter Kit for pH of Hair Care Products components

  • Orion 3-Star Benchtop pH Meter 13-642-052
  • Ross™ Ultra Semimicro Epoxy Body pH Combination Electrode 13-642-240
  • Glass Body ATC Probe 13-642-258
  • 475ml (1 qt.) each of ph 4.01, 7.00 and 10.01 buffers 13-301-133, 13-641-857, 13-079-319
  • Ross Electrode storage solution, 475ml (1 qt.) (Orion 810001)
  • Benchtop electrode stand 13-645-137
  • Stirrer and Paddle 13-641-659
  • nstruction manual and Star Series Method Note 248121-001

Dedicated Application Package for measurement of pH of Hair Care Products

Includes meter, plus all electrodes, solutions, accessories and application data required. Easy-to-use single-channel meter with Improved stability algorithm to increase calibration and measurement accuracy

Orion 3-Star™ pH Meter features:

  • Dual display of pH or mV and temperature measurements simultaneously
  • Bright LCD display with ready indication
  • Convenient built-in Quick-Start Guide
  • Automatic or manual calibration with 1 to 5 calibration points
  • Auto-Buffer Recognition (ABR) of DIN and NIST/US buffers
  • Full complement of GLP-related features
  • Stores up to 10 password protected methods, including setup parameters and calibrations; download additional methods and software updates via the Web
  • Average slope and slope of segment printed with measurement
  • New individual methods passwords
  • 1000 point datalogger, downloadable to printer or computer via an optional RS-232 or USB interface accessory; can be switched off
  • SMART STABILITY™ and SMART AVERAGING™ functions automatically optimize accuracy, precision and response time
  • ATC temperature calibration
  • Auto stirrer control; capable of operating an autosampler
  • IP54 dustproof and splashproof
  • Mini-DIN locking probe connectors
  • Advanced self-tests on meter operation, self-diagnostics
  • Universal power adapter for worldwide operation

Order Info

Additional Orion 3-Star Benchtop pH Meter Application Kits available:

  • pH of Drinking Water Water 13-643-010
  • pH of Soils 13-643-026
  • pH of Carbonated Beverages 13-643-051
  • pH of Fruit Juices 13-643-059
  • pH of Milk 13-643-045
  • pH of Processed Meats 13-643-041
  • pH of Wine and Beer 13-643-047
  • pH of Cheese 13-643-053
  • pH of Household Cleaners 13-643-067
  • pH of Hand Creams 13-643-066
  • pH of Saline Solution 13-643-073
  • pH of Soaps and Detergents 13-643-069
Additional application kits are available; contact your Fisher Scientific™ Customer Service Representative for details

Compliance

CE, CSA, UL and FCC Class A limits approved

Warranty and Services

Three-year replacement warranty

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Measures hair care product pH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.