Learn More
Thermo Scientific™ Orion™ 3-Star Benchtop Application Kit: pH of Processed Meats
Shop All Thermo Scientific ProductsDescription
Includes
Meter Kit for pH of Processed Meats components
- Orion 3-Star Benchtop pH Meter 13-642-052
- Ross™ Sure Flow pH Combination Electrode 13-642-243
- Stainless-Steel ATC probe 13-642-259
- 5 x 60mL each of ph 4.01, 7.00 and 10.01 buffers 09-357-69, 09-357-70, 09-357-68
- Electrode storage solution, 475ml (1 qt.), 13-641-908
- Benchtop electrode stand 13-645-137
- Instruction manual and Star Series Method Note 248094-001
Includes meter, plus all electrodes, solutions, accessories and application data required. Easy-to-use single-channel meter with Improved stability algorithm to increase calibration and measurement accuracy.
Orion 3-Star™ pH Meter features:
- Dual display of pH or mV and temperature measurements simultaneously
- Bright LCD display with ready indication
- Convenient built-in Quick-Start Guide
- Automatic or manual calibration with 1 to 5 calibration points
- Auto-Buffer Recognition (ABR) of DIN and NIST/US buffers
- Full complement of GLP-related features
- Stores up to 10 password protected methods, including setup parameters and calibrations; download additional methods and software updates via the Web
- Average slope and slope of segment printed with measurement
- New individual methods passwords
- 1000 point datalogger, downloadable to printer or computer via an optional RS-232 or USB interface accessory; can be switched off
- SMART STABILITY™ and SMART AVERAGING™ functions automatically optimize accuracy, precision and response time
- ATC temperature calibration
- Auto stirrer control; capable of operating an autosampler
- IP54 dustproof and splashproof
- Mini-DIN locking probe connectors
- Advanced self-tests on meter operation, self-diagnostics
- Universal power adapter for worldwide operation
Order Info
Additional Orion 3-Star Benchtop pH Meter Application Kits available:
- pH of Drinking Water 13-643-010
- pH of Soils 13-643-026
- pH of Carbonated Beverages 13-643-051
- pH of Fruit Juices 13-643-059
- pH of Milk 13-643-045
- pH of Wine and Beer 13-643-047
- pH of Cheese 13-643-053
- pH of Household Cleaners 13-643-067
- pH of Hair Care Products 13-643-068
- pH of Hand Creams 13-643-066
- pH of Saline Solution 13-643-073
- pH of Soaps and Detergents 13-643-069
Compliance
CE, CSA, UL and FCC Class A limits approved
Warranty and Services
Three-year replacement warranty
Specifications
Specifications
| For Use With (Application) | Measures processed meat pH |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.