Learn More
Thermo Scientific™ Orion™ 3-Star Benchtop pH Meter
Catalog No. 13644928 Shop All Thermo Scientific Products
Change view
Click to view available options
Orion 3-Star Plus pH Benchtop Meter Kit 1112106; includes: meter; electrode 9165BNWP; universal power supply and instructions
Catalog No.
13-644-928 Orion 3-Star Plus pH Benchtop Meter Kit 1112106; includes: meter; electrode 9165BNWP; universal power supply and instructions
1 options
Catalog No. 13-644-928 Supplier Thermo Scientific™ Supplier No. 1112106
Includes
  • Meter Only option includes meter and instruction manual.
  • Meter Kit without Electrode option includes meter, benchtop electrode stand, instruction manual.
  • Meter Kit with pH/ATC Electrode option includes meter, pH/ATC Combination Electrode 13-642-252, 475ml (1 qt.) each of ph 4.01, 7.00 and 10.01 buffers, electrode storage solution, benchtop electrode stand, instruction manual.
  • Meter Kit with Ross™ Electrode option includes meter, Ross™ Ultra™ pH Combination Electrode, Epoxy ATC probe, 475ml (1 qt.) each of ph 4.01, 7.00 and 10.01 buffers, electrode storage solution, benchtop electrode stand, instruction manual.
Dedicated measurement of pH and temperature in a compact, ergonomic package
Easy-to-use single-channel meter with improved stability algorithm to increase calibration and measurement accuracy. Reliable, accurate pH measurement for any general laboratory or QC application.
  • Displays pH or mV and temperature measurements simultaneously
  • Bright LCD display with ready indication Convenient built-in Quick-Start Guide
  • Automatic or manual calibration with 1 to 5 calibration points
  • Auto-Buffer Recognition (ABR) of DIN and NIST/US buffers
  • Offers a full complement of GLP-related features
  • Stores up to 10 password protected methods, including setup parameters and calibrations; download additional methods and software updates via the Web
  • Average slope and slope of segment printed with measurement
  • New individual methods passwords
  • 1000 point datalogger, downloadable to printer or computer via an optional RS-232 or USB interface accessory; can be switched off
  • SMART STABILITY™ and SMART AVERAGING™ functions automatically optimize accuracy, precision and response time
  • ATC temperature calibration
  • Auto stirrer control; capable of operating an autosampler
  • IP54 dustproof and splashproof
  • Mini-DIN locking probe connectors
  • Advanced self-tests on meter operation, self-diagnostics
  • Universal power adapter for worldwide operation
Warranty and Services
Three-year replacement warranty