Learn More
Thermo Scientific™ Orion™ 3-Star Plus Dissolved Oxygen Portable Meter
Shop All Thermo Scientific ProductsDescription
Includes
- Meter Only 1213000 package includes meter, batteries, and instruction manual.
- Meter Kit 1213001 package includes meter, DO probe 083010MD with 3m cable, probe service kit, DO calibration sleeve, hard field case, batteries, and instruction manual.
- Meter Kit 1213101 package includes meter, DO probe 083010MD with 3m cable, probe service kit, DO calibration sleeve, batteries, and instruction manual.
- Meter Kit 1213501 package includes meter, DO probe 083010MD with 3m cable, probe service kit, DO calibration sleeve, soft field case, batteries, and instruction manual.
Delivers over 2000 hours of continuous operation, with improved stability algorithm to increase calibration and measurement accuracy. Easy to set up and use, provides reliable measurements for water testing applications.
Easy to operate in any mode
- Bright, backlit LCD display with ready indication Balanced comfortable handling
- Simultaneous display of DO and temperature
- Convenient built-in Quick-Start Guide with calibration instructions
- Stores, displays, and prints up to 10 methods with individual method password protection
- Automatic temperature compensation to two decimal places
- Auto-read/hold function
- IP67 waterproof
- Automatic barometric pressure and salinity compensation
- Calibration to Winkler Titration for BOD measurements
- Solution and membrane temperature printed and displayed with measurement
- Calibrate both solution and membrane temperature
- Uses polarographic DO probe
Increased accuracy
- Improved stability algorithm improves calibration and measurement accuracy
- SMART STABILITY™ and SMART AVERAGING™ functions optimize accuracy, precision, and response time
- ATC calibration
- Advanced self-tests on meter operation, self-diagnostics
- Full complement of GLP-related features
- 1000 point datalogger, downloadable to printer or computer via RS-232 connection
- Average slope and slope of segment printed with measurement
Certifications
CE, CSA, UL, TUV and FCC Class A limits approved
Compliance
21 CFR Part II Compliance with optional software
Warranty and Services
Three years