Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ 55PR Printers

Catalog No. 13301129 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
pH3695-5/-7, 55PR40/49 Printer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-301-129 pH3695-5/-7, 55PR40/49 Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-301-129 Supplier Thermo Scientific™ Supplier No. 55PR45

missing translation for 'validMainframeMsg'

Specifications

Type Printer Paper
For Use With (Application) pH
For Use With (Equipment) pH3695-5/-7, 55PR40/49 Printer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.