Learn More
Thermo Scientific™ Orion™ Fruit Juices Meter Test Kits
Catalog No. 13643058 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Calcium
Potassium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
13-643-058 Potassium
13-643-057 Calcium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-643-058 Supplier Thermo Scientific™ Supplier No. 1010150
Specifications
For Use With (Application) Potassium
Includes 4-star benchtop pH/ISE meter, ionplus potassium electrode, standard (0.1MKcl), potassium ISA, 475mL, optimum results E reference fill solution
Product Type Fruit Juices Meter Test Kit