Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Green pH Combination Electrodes
SCI_ED

Catalog No. S04956 Shop All Thermo Scientific Products
Change view
Click to view available options
Junction:
Double
Temperature (Metric) Range:
0°C to 80°C
0°C to 90°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Junction Temperature (Metric) Range
S04956 Double 0°C to 90°C
S04970 Double 0°C to 80°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04956 Supplier Thermo Scientific™ Supplier No. GD9156BNWP/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Available in low maintenance gel versions for simplified preparation and use or refillable version for improved longevity.

  • RoHS compliant—no lead, mercury or other hazardous materials
  • No disposal issues, no disposal cost
  • Choice of single-junction models for routine sample measurements, or double-junction models for biological samples or samples that contain Tris or sulfides
  • Choice of refillable or low-maintenance gel-filled models
  • Duarable epoxy body
  • pH Range: 0 to 14
  • Dimensions: 120 x 12mm, cap dia. 16mm
  • Waterproof BNC connector, 1m cable; fit any meter with BNC input
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC (Waterproof)
Junction Double
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Type Double Junction pH Electrode
For Use With (Equipment) Orion Star Series Meters
Temperature (Metric) Range 0°C to 90°C
Length (English) Cable 39.3 in.
Length (Metric) Cable 1 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.