Learn More
Thermo Scientific™ Orion™ Maintenance Kits, Membranes and Electrolytes
Shop All Thermo Scientific ProductsSpecifications
Specifications
| For Use With (Equipment) | 820, 840, 860, 870 and 880 Series Galvanic Probes |
| Type | Probe Maintenance Kit |
| Includes | Electrolyte Solution, cleaning solution, cleaning paper |