Learn More
Thermo Scientific™ Orion™ Milk Meter Test Kits
Specifications
Specifications
| For Use With (Application) | Calcium |
| Includes | 4-star benchtop pH/ISE meter, ionplus (R) calcium electrode with waterproof BNC connector, standard, calcium ISA, 475mL, optimum results A reference fill solution |
| Product Type | Milk Meter Test Kit |