Learn More
Thermo Scientific™ Orion™ Processed Meats Meter Test Kits
Catalog No. 13643039
Change view
Click to view available options
For Use With (Application):
Nitrate
Nitrite
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
13-643-039 Nitrate
13-643-040 Nitrite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-643-039 Supplier Thermo Scientific™ Supplier No. 1010131
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Specifications
For Use With (Application) Nitrate
Includes 4-star benchtop pH/ISE meter, nitrate half cell, ionplus double junction electrode, standard, optimum results F, ISA, internal fill, interference suppressor
Product Type Processed Meats Meter Test Kit