Learn More
Thermo Scientific™ Orion™ Series 200 Portable Multiparameter Meters
Catalog No. 13641248 Shop All Thermo Scientific Products
Change view
Click to view available options
With electrode
Catalog No.
13-641-248 With electrode
1 options
Catalog No. 13-641-248 Supplier Thermo Scientific™ Supplier No. 0290A0
True one-handed operation—ideal for field measurements. Feature splash-resistant sealed housing with tactile/audible keypad, improved cable routing, and wet storage sleeve for electrode to prevent dry out. A pop-out stand on the back lets you use them on a benchtop. All feature large LCD with simultaneous display of pH or ISE measurement and temperature. User prompts, stability indicator, diagnostics and 80 to 120% slope control make meters reliable and easy to use. Selectable features include autohold, to lock in stable readings; beep, to confirm stable data points; auto shutoff, to conserve batteries without data loss; and manual or auto calibration and temperature compensation. All models have ORP Mode that allows standardization to the Standard or Normal Hydrogen Electrode used to define Oxidation Reduction Potential (ORP).
Meter/Electrode packaging for Models 290Aplus, 250Aplus and 230Aplus includes the Orion Triode™ Combination pH Electrode/ATC Probe (13-642-545) to eliminate need for separate ATC probe. All come with 9V battery (AC adapter is optional and sold separately) and instruction manual. Models 250Aplus and 230Aplus are also available in Meter/Starter Kit packaging, including 13-642-545 Triode Combination Electrode/ATC Probe, battery, and instruction manual; electrode holder and stand; two 2 oz. (60mL) bottles each of pH 4, 7, and 10 buffers; filling solution; beaker; two electrode clips; and fitted carrying case. (Starter Kit without meter and electrode also available separately; contact your Fisher Customer Service Representative) Model 250Aplus is also available with Meter/AquaPro™ pH Kit containing 13-641-217 AquaPro Electrode, 13-641-1119 ATC probe, buffers, beaker, stand, electrode holder and field case.
Orion 290 Aplus™ pH/ISE Meter offers one- to five-point pH autocalibration with built-in recognition for five buffers; manual ISE calibration up to five points with autoblank correction; selectable resolution of 0.1/0.01/0.001pH; autocalculation/display of electrode slope; adjustable isopotential point in pH; and time/date of last calibration displayed. Has 24-hour clock, interval timer and high/low alarm limits; and internal 25-point datalogging. Three print options: when signal is stable, at timed interval, or on demand. Two-way RS-232-C port transfers data to either printer (13-641-257, sold separately) or computer.
Orion 250Aplus pH/mV Meter features three-point pH autocalibration, built-in recognition of three buffers; selectable resolution of 0.1/0.01pH; autocalculation/display of electrode slope; adjustable isopotential point; and time/date of last calibration displayed. Has 24-hour clock, interval timer and datalogging capability. Three print options: when signal is stable; at timed interval; on demand. RS-232-C output.
Orion 230Aplus pH/mV Meter performs two-point pH autocalibration, recognizes three buffers, calculates and displays electrode slope values, and offers simultaneous display of pH and temperature.
Warranty and Services
Three-year warranty
Key to Special Features:A. Custom LCD (liquid crystal display).B. Auto blank correction with three or more ISE points.C. Autocalibrates with two pH buffers.D. Autocalibrates with three pH buffers.E. Autocalibrates with five pH buffers.F. Timer; high/low alarm limits.G. Datalogging capability.H. User-friendly prompts and self-diagnosis.I. 3 print options: when electrode signal is stable, at selected timer interval, or on demand.J. Works with optional Model 900A printer.
Footnotes:
† Selectable to 0.1, 0.01 or 0.001pH.
† Relative mV range: 0 to ± 1999.9mV.