Learn More
Thermo Scientific™ Orion™ Turbo2™ Balance Interface Cables
Catalog No. 13642730
Change view
Click to view available options
Cable F/balances
Cable Kf To Mettler Ae Balance
Cable Kf To Mettler At Balance
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-642-730 Cable F/balances
13-642-732 Cable Kf To Mettler Ae Balance
13-642-735 Cable Kf To Mettler At Balance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-642-730 Supplier Thermo Scientific™ Supplier No. 319035038
Enable connection of Turbo2, AF8, AF7 and AF7LC titrators to balances listed. The Cahn™/Thermo Orion CO-10 balance requires 13-642-730 Cable.