Learn More
Thermo Scientific™ Orion™ Wastewater Meter Test Kits
Catalog No. 13643022 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
ORP
Sulfide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
13-643-022 Sulfide
13-643-021 ORP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-643-022 Supplier Thermo Scientific™ Supplier No. 1010113
Specifications
For Use With (Application) Sulfide
Includes Orion 5-Star Benchtop Multimeter, Sure-Flow Combination Silver/Sulfide Electrode, SAOB reagent
Product Type Wastwater Meter Test Kit