Learn More
Thermo Scientific™ Owl™ TBE Buffer
Catalog No. OWER24S Shop All Thermo Scientific Products
Catalog No. OW-ER24-S Supplier Thermo Scientific™ Supplier No. ER24S
Nucleic acid electrophoresis running buffer for PAGE and agarose gels
  • Prepackaged powdered buffer: Tris 89mM; Boric acid 89mM; EDTA 2.6mM, pH 8.3