Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Influenza A & B
Catalog No. RK6105112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
RK6105112 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. RK6105112 Supplier Thermo Scientific™ Supplier No. K6105112
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the detection and identification of Influenza virus strains A and B in clinical specimens or cell cultures
  • Test utilizes species-specific monoclonal antibodies to detect epitopes of Influenza virus glycoproteins and fusion proteins specific to either Influenza virus type A or Influenza virus type B
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 Tests