Learn More
Thermo Scientific™ Tripod Stand for PackEye Radiation Detection Backpack
GSA_VA
Catalog No. 19175641 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Tripod
Catalog No. Product Type
19-175-641 Tripod
1 options
Catalog No. 19-175-641 Supplier Thermo Scientific™ Supplier No. 4255086
Add to Cart
Add to Cart
Tripod Stand for use with FHT 1377 GN-2 PackEye Radiation Detection Backpack.
  • Easily adjustable tripod stand with mounting plate to hold the PackEye case (not included)
  • For use with FHT 1377 GN-2 PackEye Radiation Detection Backpack
  • The PackEye system was developed for the rapid detection and location of gamma emitting radioactive sources
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tripod