Learn More
Thermo Scientific™ Thermo Scientific™ PCR Capping Tool
Catalog No. A55327 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
SureTect assays
Catalog No. For Use With (Equipment)
A55327 SureTect assays
1 options
This item is not returnable. View return policy
Catalog No. A55327 Supplier Thermo Scientific™ Supplier No. A55327
Add to Cart
Add to Cart
This item is not returnable. View return policy
Easily and quickly cap both the lysis and assay tubes in the Thermo Scientific™ SureTect™ PCR Assay workflow using the Thermo Scientific ™PCR Capping Tool.
Easily and quickly cap both the lysis and PCR tubes in the Thermo Scientific™ SureTect™ PCR Assay workflow using the Thermo Scientific™ PCR Capping Tool.
Designed specifically for use with the SureTect assays, the Thermo Scientific™ Capping Tool is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific PCR Capping Tool to quickly and easily apply caps to the SureTect lysis and PCR plates guaranteeing a tight seal in a fast and easy to use workflow
Quick and Easy
Use the Thermo Scientific PCR Capping Tools to speed the process of capping multiple lysis and assay strips. This tool is designed according to the tube and cap dimensions so you can quickly cap lysis and PCR tube strips. The rugged material and design endures constant use so can be used time and time again whatever your sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR Workflow is continually evolving to support your laboratory and testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Description Thermo Scientific™ PCR Capping Tool
Type Capping Tool
For Use With (Equipment) SureTect assays