Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ 6 Protein Digest, equimolar, LC-MS grade

Catalog No. PI88342 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 pmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88342 100 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88342 Supplier Thermo Scientific™ Supplier No. 88342
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Calibrate and maintain quality control for mass spectrometry (MS) analysis of proteomic samples using the Thermo Scientific™ Pierce™ 6 Protein Digest Standard, Equimolar, LC-MS Grade, a highly validated equimolar protein digest mixture.

The Thermo Scientific Pierce 6 Protein Digest Standard, Equimolar, LC-MS Grade, is a highly validated equimolar protein digest mixture for use as a quality control sample for mass spectrometry (MS) analysis of proteomic samples.

  • Positive control sample - optimized and validated as a quality control standard for mass spectrometry applications
  • Excellent sequence coverage - on average, greater than 70% sequence coverage of the 6-protein mixture
  • Validated peptide quality - digestion procedure optimized for minimal missed cleavages and over-alkylation
  • Rigorously tested - high-quality, consistent, 6-protein digest guaranteed and documented via lot-specific certificates of analysis
  • Stable - provided in a stable lyophilized format
The Pierce 6 Protein Digest Standard contains an equimolar mixture of highly pure bovine cytochrome c, lysozyme, alcohol dehydrogenase, bovine serum albumin, apo-transferrin, and beta-galactosidase. The proteins are reduced, alkylated (iodoacetic acid), digested (MS-grade trypsin), and desalted (C18) after trypsin digestion to provide a quality LC/MS-grade standard free of intact protein. The peptide digest provides excellent sequence coverage with minimal over-alkylation and missed cleavages. This digest has been robustly tested for peptide quality, digestion efficiency, and lot-to-lot uniformity and is suitable for LC and LC-MS applications.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Protein Digest
Product Line Pierce
Content And Storage Store in freezer (-5 to -30°C).
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 100 pmol
Form Solid
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.