Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monosodium Salt, 1 g
GSA_VA

Catalog No. PI88292 Shop All Thermo Scientific Products
Change view
Click to view available options
Monosodium Salt; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI88292 Monosodium Salt; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88292 Supplier Thermo Scientific™ Supplier No. 88292
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.

Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.

We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C.
Detection Method Bioluminescence
For Use With (Application) ATP Assay, Gene Reporter Assay
Product Type D-Luciferin, Monosodium Salt
Product Line Pierce
Quantity 1 g
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.