Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNase I Solution (1 unit/μL), RNase-free

Catalog No. PI89836 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89836 1000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89836 Supplier Thermo Scientific™ Supplier No. 89836
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleave and degrade unwanted single- and double-stranded DNA

Deoxyribonuclease I (DNase I) is a single, glycosylated polypeptide that degrades unwanted single- and double-stranded DNA. The enzyme works by cleaving DNA into 5' phosphodinucleotide and small oligonucleotide fragments. DNase I is commonly added to cell lysis reagents to remove the viscosity caused by the DNA content in bacterial cell lysates or to remove the DNA templates from RNAs produced by in vitro transcription. Two grades of DNAse are offered, one sufficient for protein work and one that is useful for any application requiring the digestion of DNA in which it is crucial to avoid damage to RNA.

Highlights:

  • Degrades and removes unwanted DNA from samples
  • Cleaves both single-stranded and double-stranded DNA
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • Reduces viscosity of bacterial lysates (protein extracts) to facilitate pipetting
  • Supplied as 50% glycerol stocks in 10mM Tris-HCl pH 7.5, 10mM CaCl2, 10mM MgCl2
  • Available in two convenient formats: RNase-tested to protect RNA (Part No. 89836) and convenient protein extraction grade (Part No. 90083)
General information about the use of DNase I:
  • Calcium ions are required for DNase I activity, and trace amounts of Ca++ may be present at high enough concentrations for DNase I to be active; however, the use of EGTA or calcium-free buffers can reduce DNase I activity to undetectable levels
  • High levels (i.e., 100mM) of monovalent ions such as Na+ and K+ will decrease DNase I activity
  • DNase I is inactivated by heating to 65°C for 10 minutes
  • Kunitz unit: 1 Kunitz unit is the amount of enzyme required to cause an increase in absorbance at 260nm of 0.001/min/mL at 25°C in 0.1 M NaOAc, pH 5.0, due to the degradation of highly polymerized DNA
  • Degradation assay units: 1 unit is defined as the amount of enzyme required to completely degrade 1mg of plasmid DNA in 10 minutes at 37°C in 10mM Tris-HCl, pH 7.5, 50mM MgCl2, 13mM CaCl2

Specifications (Part No. 89836):
  • Visual: Clear liquid
  • Concentration: 1 Unit/mL
  • Activity: ≥ 100,000 Units/mg protein
  • RNase Contamination: 20 Units release < 2% non-ethanol precipitable when incubated with 3H-labeled Poly(A) RNA
  • Functional Analysis: 1μL enzyme degrades < 5% RNA but degrades 100% DNA

Specifications (Part No. 90083):
  • Visual: Clear, colorless liquid, free of insoluble material
  • Units: ≥2500 units/mL
  • Unit definition: 1 unit is defined as the amount of enzyme required to produce an increase in absorbance at 260nm of 0.001/min/mL at 25°C of highly polymerized DNA

 

 

  • Elimination of viscosity caused by DNA in protein cell lysates
  • Removal of DNA templates from RNAs produced by in vitro transcription

 

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Store at -20°C
Form Liquid
Enzyme DNase
Quantity 1000 Units
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.