Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Ninhydrin (2,2-dihydroxy-1H-indene-1,3(2H)-dione)

Catalog No. pi21003
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21003 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21003 Supplier Thermo Scientific™ Supplier No. 21003BID
Only null left
Add to Cart
Add to Cart

Label, detect and measure peptides, proteins and amino acids by HPLC (liquid chromatography) using this amine-reactive, colored reagent (Amax 440nm).

Thermo Scientific™ Pierce Ninhydrin is a classic, amine-reactive, colorimetric detection reagent for protein detection and amino acid analysis.

Highlights:

  • Used in amino acid chromatography
  • Offers superb color response and low blank

Ordering Alerts:

Indefinitely stable. No refrigeration required. Keep bottle tightly sealed. Avoid exposure to direct sunlight and ammonia.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Ninhydrin
Chemical Reactivity Amine
Product Line Pierce
Content And Storage Store in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Label or Dye Other Label or Dye
Detection Method Colorimetric
Quantity 500 g

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.