Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Poly-HRP Streptavidin

Catalog No. ENN200 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN200 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ENN200 Supplier Thermo Scientific™ Supplier No. N200
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An advanced polymer of HRP conjugated to streptavidin with a high HRP:streptavidin ratio

  • Produces an enhanced signal
  • May increase ELISA sensitivity
  • Validated for use with some Thermo Scientific Pierce Matched Antibody Pairs and MiniKits

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0 mg/mL
Conjugate HRP
Quantity 0.25 mL
Content And Storage Do not store below -20°C. Avoid multiple freeze-thaw cycles.
Form Liquid
Product Type Streptavidin Conjugate
Target Molecule Biotin
Label or Dye HRP (Horseradish Peroxidase)
What is the concentration of Poly-HRP Streptavidin (Cat. No. N200) and what is the labelling ratio of HRP:Streptavidin?

The concentration of Poly-HRP Streptavidin is 1 mg/mL. The HRP:Streptavidin ratio is 5:1.

Can Poly-HRP Streptavidin (Cat. No. N200) be stored at -80 degrees C to increase its shelf life?

We do not recommend storing Poly-HRP Streptavidin at -80 degrees C, as the product can freeze solid at temperatures below -25 degrees C even if it has 50% glycerol as a stabilizer, thereby necessitating thawing of the product. With multiple freeze-thaws, the enzyme conjugates are rendered unstable.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.