Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Neuronal Culture Medium for Pierce™ Primary Cell Isolation Kits

Catalog No. PI88283 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88283 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88283 Supplier Thermo Scientific™ Supplier No. 88283
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Easily isolate and culture highly viable functional primary neurons

The Thermo Scientific Pierce Primary Neuron Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary neurons form embryonic cortical and hippocampal tissue from mouse and rat.

The Primary Neuron Isolation Kit has been designed to obtain high viability and purity of primary embryonic neurons using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains tissue-specific dissociation enzymes, media formulated specifically for neuronal cell culture, and cell culture supplements that promote the isolation and growth of functional neurons. The validated protocol and reagents produce highly pure primary neurons for either short- or long-term culture, and the kit is designed for use by both experienced and non-experienced users.

Highlights:

  • Optimized - procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Yield - provides a 2-fold increase in yield compared to do-it-yourself methods
  • Viability - greater than 95% of isolated cells remain viable in culture from Day 1 to Week 4
  • Functional - cultured neurons contain processes indicative of complete differentiation and functionality
Includes:
  • Complete kit includes Neuronal Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), Neuronal Culture Medium, Neuronal Culture Media Supplement, Glutamine Supplement, and Neuronal Growth Supplement
Requires:
  • Fresh embryonic mouse or rat tissue sample, heat-inactivated fetal bovine serum (FBS), poly-D-lysine treated culture dishes
Applications:
  • Neuronal cell differentiation
  • Immunohistochemistry (IHC)
  • Functional and biochemical assays
  • Biologically relevant system for preclinical drug discovery, neurotoxicity testing, and predictive disease modeling
Product Details:
The Pierce Primary Neuron Isolation Kit has been optimized to provide excellent cell yield and to sustain cells at greater than 95% viability. The neurons are appropriately polarized, develop extensive axonal and dendritic arbors, express neuronal and synaptic markers, and form numerous, functional synaptic connections with one another. They can be used as a model system for molecular and cellular biology studies of neuronal development and function, especially for visualizing the subcellular localization of endogenous or expressed proteins, imaging protein, neuronal polarity, and dendritic growth and synapse formation [1].
General References:
1. Silva, R.F.M, et al. (2006) Dissociated primary nerve cell cultures as models for assessment of neurotoxicity. Toxicology Letters 163:1-9.
2. Ivenshitz, M. and Segal, M, (2009). Neuronal density determines network connectivity and spontaneous activity in cultured hippocampus. J Neurophysiol 104:1052-60.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primary Cell Isolation Media
Content And Storage Store at 4°C, Protect from light
Cell Type Neuronal Cells (Pre-Natal), Primary Cells
Target Species Rat, Mouse
Form Liquid
Product Line Pierce
For Use With (Application) Immunohistochemistry
Quantity 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.