Learn More
Description
A broad range of pocket volumes are compatible with many industry standard 96- and 384-channel automated liquid handling platforms to allow existing equipment to implement new innovative pipetting procedures.
- Transfer samples directly to the assay plate
- Combine sample and assay material with soft in-tip mixing—thorough mixing without damaging sensitive material
- Consistent accuracy and precision; coefficient of variation of less than 6% can be achieved for any pocket size
- Color: Clear
Specifications
Specifications
| Volume (Metric) | 250 nL |
| Sterility | Sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | PerkinElmer, Hamilton MICROLAB STAR |
| Product Line | PocketTip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.