Learn More
Thermo Scientific™ RadEye™ PRD-ER High-Sensitivity Personal Radiation Detectors
Catalog No. 19173336
Change view
Click to view available options
Factory calibrated in exposure rate R/h; Black
Catalog No.
19-173-336 Factory calibrated in exposure rate R/h; Black
1 options
Catalog No. 19-173-336 Supplier Thermo Scientific™ Supplier No. 425067104
Ideal interdiction and response tool features special proprietary circuitry allows energy-compensated dose and dose rate measurement up to 100mSv/h (or 10 rem/h).
Advanced pocket-size instrument detects and localizes radiation sources as generated by man-made devices such as nuclear weapons, improvised nuclear devices (IND) or radiological dispersal devices (RDDs).
Single detector arrangement offers the following unique advantages over the whole measuring range:
  • Consistent angular dependence
  • No mutual shielding of neighbored detectors
  • Consistent energy response
  • No transition range with annyong hysteresis effects
  • No high activity source for function test of high dose rate detector required
  • Menu-driven with an intuitive, easy-to-use format
  • Patented NaturalBackground Rejection (NBR) technology provides high sensitivity withhigh selectivity; eliminates nuisance alarming due to naturallyrecurring radiation sources (such as granite)
  • Low-power-technology components and fully automated self-checks result in minimum maintenance
  • Equipped with large clear graphic display and audible/visible/vibrating alarms; earphone-output for silent alarming