Learn More
Thermo Scientific™ RAP Regulated Air Pumps
GSA_VA
Catalog No. 19175534 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Collecting sampling media onto filters, pulling airborne gases through active sampling systems,or radiological detection applications
Catalog No. For Use With (Application)
19-175-534 Collecting sampling media onto filters, pulling airborne gases through active sampling systems,or radiological detection applications
1 options
Catalog No. 19-175-534 Supplier Thermo Scientific™ Supplier No. RAP1
Add to Cart
Add to Cart
Compact, portable system containing an oilless vacuum pump, motor and airflow regulator ideal for collecting sampling media onto filters, or pulling airborne gases through active sampling systems, for radiological detection applications
Designed to maintain a constant pressure drop across an in-line orifice by controlling a variable bypass valve into the pump.
  • Durable construction
  • Adjustable orifice for flow rate adjustment from near zero up to the maximum pump flow capacity
  • Flow control system permits the pump to operate at a minimum pressure drop at all times which provides cooler pump operation to extend the lifetime
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Collecting sampling media onto filters, pulling airborne gases through active sampling systems,or radiological detection applications
Includes Oilless vacuum pump, motor and airflow regulator