Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Anaerobic Reducible CNA Blood Agar/ Kanamycin Bile Esculin Agar
Click to view available options
:
Anaerobic Reducible CNA Blood Agar
Specifications
Specifications
| Certifications/Compliance | USDA, APHIS |
| Manufacturing Origin | United States |
| Media Type | Anaerobic reducible CNA Blood Agar/Kanamycin Bile Esculin Agar |
| Formulation | Kanamycin Sulphate <0.1%, Nalidixic Acid <0.1% |
| For Use With (Application) | Isolation and differentiation of microorganisms from samples or specimens |
| Packaging Type | Biplate |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction